Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013353.2 | NP_037485.2 | 6 | 743 | ||
XM_011509449.1 | XP_011507751.1 | 6 | 625 | |||
XM_017001089.1 | XP_016856578.1 | 5 | 1360 | |||
XM_017001090.1 | XP_016856579.1 | 6 | 672 | |||
GenBank mRNA | AF165217.1 | - | 6 | 620 | ||
AF177173.2 | - | 6 | 688 | |||
AK092700.1 | - | 6 | 609 | |||
AK300675.1 | - | 4 | 446 | |||
AK313280.1 | - | 6 | 652 | |||
BC017810.1 | - | 6 | 653 | 4288769 | ||
BX647346.1 | - | 5 | 475 |