Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF078859.1 | - | 6 | 726 | ||
AF078868.1 | - | 6 | 707 | |||
AF116703.2 | - | 3 | 284 | |||
AF134478.1 | - | 6 | 682 | |||
AK027523.1 | - | 6 | 759 | |||
AK074710.1 | - | 5 | 657 | |||
AK225112.1 | - | 6 | 695 | |||
AK225138.1 | - | 6 | 735 | |||
AK296339.1 | - | 6 | 675 | |||
AL136546.1 | - | 5 | 614 | |||
AL833441.1 | - | 6 | 723 | |||
BC012842.1 | - | 6 | 796 | 3861915 | ||
BC013925.1 | - | 6 | 638 | 3685016 | ||
BC029376.1 | - | 6 | 675 | 4658276 | ||
BC091522.1 | - | 6 | 650 | 30513213 | ||
DQ250006.1 | - | 6 | 651 |