Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013286.4 | NP_037418.3 | 1 | 2276 | ||
GenBank mRNA | AL831838.1 | - | 1 | 1682 | ||
AY545557.1 | - | 1 | 250 | |||
BC001367.2 | - | 1 | 819 | |||
BC121184.2 | - | 2 | 157 | |||
BC139836.1 | - | 2 | 1487 | 40123447 | ||
L13434.1 | - | 1 | 2033 |