Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271681.1 | NP_001258610.1 | 3 | 383 | ||
NM_013328.3 | NP_037460.2 | 3 | 383 | |||
GenBank mRNA | AF087859.1 | - | 3 | 301 | ||
AF151351.1 | - | 3 | 181 | |||
AK001508.1 | - | 2 | 1638 | |||
AK291913.1 | - | 3 | 218 | |||
AL834217.1 | - | 1 | 380 | |||
BC014868.1 | - | 3 | 274 | 3919161 | ||
BC020553.1 | - | 3 | 308 | 2901360 | ||
BU538765.1 | - | 3 | 258 | |||
HY044355.1 | - | 3 | 384 | |||
KU178615.1 | - | 3 | 153 |