Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013330.4 | NP_037462.1 | 9 | 1094 | ||
NM_197972.2 | NP_932076.1 | 9 | 1063 | |||
NR_104229.1 | 9 | 1094 | ||||
XM_005245106.4 | XP_005245163.1 | 7 | 672 | |||
XM_017001099.1 | XP_016856588.1 | 9 | 1080 | |||
XM_017001100.1 | XP_016856589.1 | 7 | 660 | |||
GenBank mRNA | AF153191.2 | - | 9 | 933 | ||
AK094513.1 | - | 9 | 1046 | |||
AK290701.1 | - | 9 | 928 | |||
AK301917.1 | - | 9 | 1077 | |||
BC006983.1 | - | 9 | 925 | 3997152 |