Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256185.1 | NP_001243114.1 | 3 | 346 | ||
NM_001256186.1 | NP_001243115.1 | 2 | 310 | |||
NM_001256187.1 | NP_001243116.1 | 2 | 322 | |||
NM_001256188.1 | NP_001243117.1 | 2 | 310 | |||
NM_013346.3 | NP_037478.2 | 2 | 322 | |||
GenBank mRNA | AF171229.1 | - | 2 | 199 | ||
AK312977.1 | - | 2 | 270 | |||
BC020559.1 | - | 2 | 214 | |||
BC080535.1 | - | 2 | 197 | |||
BC103847.2 | - | 2 | 212 | 40002749 | ||
BC103848.1 | - | 2 | 212 | 40002752 | ||
BM805683.1 | - | 369 | ||||
BT007203.1 | - | 2 | 180 | |||
DA439434.1 | - | 2 | 258 | |||
DA736880.1 | - | 3 | 346 | |||
DA847821.1 | - | 2 | 263 | |||
DB251077.1 | - | 2 | 278 | |||
DB482683.1 | - | 2 | 322 |