Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080976.2 | NP_001074445.1 | 6 | 2906 | ||
NM_001322937.1 | NP_001309866.1 | 7 | 3462 | |||
NM_001322938.1 | NP_001309867.1 | 7 | 3374 | |||
NM_001322939.1 | NP_001309868.1 | 6 | 2789 | |||
NM_001322940.1 | NP_001309869.1 | 7 | 3458 | |||
NM_001322941.1 | NP_001309870.1 | 6 | 2956 | |||
NM_001322943.1 | NP_001309872.1 | 4 | 2512 | |||
NM_001322944.1 | NP_001309873.1 | 5 | 2752 | |||
NM_013352.3 | NP_037484.1 | 6 | 2960 | |||
NR_136524.1 | 5 | 2694 | ||||
XM_017010795.1 | XP_016866284.1 | 8 | 3095 | |||
XM_017010796.1 | XP_016866285.1 | 5 | 2716 | |||
XM_017010797.1 | XP_016866286.1 | 5 | 3432 | |||
GenBank mRNA | AF098066.1 | - | 6 | 2804 | ||
AK128607.1 | - | 2 | 2558 | |||
AK301513.1 | - | 6 | 2789 | |||
AL832471.1 | - | 1 | 2056 | |||
BC039245.1 | - | 6 | 2851 | 5272885 |