Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013371.3 | NP_037503.2 | 4, 5 | 1145 | ||
NM_153758.2 | NP_715639.1 | 3, 4 | 345 | |||
XM_011509450.2 | XP_011507752.1 | 3, 4 | 784 | |||
GenBank mRNA | AF192498.1 | - | 3, 4 | 253 | ||
AF453946.1 | - | 4, 5 | 1145 | |||
AY040367.1 | - | 3, 4 | 345 | |||
FJ380053.1 | - | 2, 3 | 206 |