Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDFIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166345.1 | NP_001159817.1 | 4 | 1034 | ||
NM_199072.4 | NP_951038.1 | 4 | 1034 | |||
GenBank mRNA | AF054589.1 | - | 4 | 1034 | ||
AY196485.1 | - | 3 | 528 |