Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BAZ2B SIRNA Assays ›
Gene Symbol:
BAZ2B
Gene Name:
bromodomain adjacent to zinc finger domain 2B
Gene Aliases:
WALp4
Chromosome Location:
Chr.2: 159315312 - 159712439 on Build GRCh38
UniGene ID:
Hs.470369
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26866
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BAZ2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289975.1
NP_001276904.1
26
4407
NM_013450.3
NP_038478.2
27
4515
XM_005246488.2
XP_005246545.2
28
4391
XM_005246489.4
XP_005246546.2
27
5494
XM_005246492.4
XP_005246549.2
27
5489
XM_006712461.3
XP_006712524.2
31
4965
XM_011511038.2
XP_011509340.1
28
4489
XM_011511039.1
XP_011509341.1
25
4112
XM_011511042.1
XP_011509344.1
26
4145
XM_011511043.1
XP_011509345.1
24
4010
XM_011511044.1
XP_011509346.1
25
4037
XM_011511045.1
XP_011509347.1
25
3950
XM_011511047.1
XP_011509349.1
27
4297
XM_011511048.2
XP_011509350.1
27
4362
XM_011511050.1
XP_011509352.1
25
3845
XM_011511051.1
XP_011509353.1
25
4031
XM_011511052.1
XP_011509354.1
24
3749
XM_011511053.2
XP_011509355.1
26
4145
XM_011511054.2
XP_011509356.1
26
4145
XM_011511056.1
XP_011509358.1
16
1765
XM_017003918.1
XP_016859407.1
31
4955
XM_017003919.1
XP_016859408.1
31
4949
XM_017003920.1
XP_016859409.1
33
5166
XM_017003921.1
XP_016859410.1
28
4489
XM_017003922.1
XP_016859411.1
28
4385
XM_017003923.1
XP_016859412.1
26
4139
XM_017003924.1
XP_016859413.1
25
4037
XM_017003925.1
XP_016859414.1
27
4393
XM_017003926.1
XP_016859415.1
27
4251
XM_017003927.1
XP_016859416.1
27
4291
XM_017003928.1
XP_016859417.1
27
4201
XM_017003929.1
XP_016859418.1
27
4283
XM_017003930.1
XP_016859419.1
27
4091
XM_017003931.1
XP_016859420.1
27
4189
XM_017003932.1
XP_016859421.1
24
3842
XM_017003933.1
XP_016859422.1
26
4068
XM_017003934.1
XP_016859423.1
26
3963
XM_017003935.1
XP_016859424.1
25
3901
XM_017003936.1
XP_016859425.1
25
3855
XM_017003937.1
XP_016859426.1
20
2562
GenBank mRNA
AB032255.1
-
22
3888
AB040909.2
-
26
4375
AK027612.1
-
18
2142
AK074895.1
-
5
633
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0