Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLTSCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015710.4 | NP_056525.2 | 3 | 337 | ||
GenBank mRNA | AF182076.1 | - | 3 | 320 | ||
AF296124.1 | - | 3 | 289 | |||
AK024486.1 | - | 3 | 308 | |||
AK225915.1 | - | 3 | 366 | |||
AK297877.1 | - | 3 | 323 | |||
AK301117.1 | - | 3 | 327 | |||
AK303918.1 | - | 324 | ||||
AK309788.1 | - | 3 | 325 | |||
AL359335.1 | - | 3 | 256 | |||
AL359336.1 | - | 3 | 280 | |||
AY535000.1 | - | 3 | 313 | |||
BC004229.2 | - | 3 | 297 | |||
BC006311.2 | - | 3 | 319 | 4111480 | ||
BC007248.1 | - | 3 | 301 | |||
BC010095.2 | - | 3 | 356 | 3501649 | ||
BC013203.1 | - | 3 | 325 | |||
BC013235.1 | - | 3 | 308 | |||
BC013307.1 | - | 3 | 309 | |||
BC014009.2 | - | 3 | 307 | |||
BC039869.1 | - | 3 | 309 | |||
CX866974.1 | - | 3 | 337 |
Gene Symbol: LOC440311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077061.1 | 1 | 423 | |||
GenBank mRNA | AW103269.1 | - | 1 | 388 |