Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASPH SIRNA Assays ›
Gene Symbol:
ASPH
Gene Name:
aspartate beta-hydroxylase
Gene Aliases:
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location:
Chr.8: 61500556 - 61714640 on Build GRCh38
UniGene ID:
Hs.332422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s269
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASPH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164750.1
NP_001158222.1
2
252
NM_001164751.1
NP_001158223.1
2
252
NM_001164752.1
NP_001158224.1
2
252
NM_001164753.1
NP_001158225.1
2
252
NM_001164754.1
NP_001158226.1
2
447
NM_001164755.1
NP_001158227.1
2
447
NM_001164756.1
NP_001158228.1
2
447
NM_004318.3
NP_004309.2
2
447
NM_020164.4
NP_064549.1
2
252
NM_032466.3
NP_115855.1
2
447
NM_032467.3
NP_115856.1
2
252
NM_032468.4
NP_115857.1
2
252
XM_005251235.2
XP_005251292.1
2
416
XM_005251236.2
XP_005251293.1
2
416
XM_005251238.2
XP_005251295.1
2
416
XM_005251239.2
XP_005251296.1
2
416
XM_005251240.1
XP_005251297.1
2
194
XM_005251242.2
XP_005251299.1
2
416
XM_005251243.2
XP_005251300.1
2
416
XM_005251244.1
XP_005251301.1
2
194
XM_005251246.2
XP_005251303.1
2
416
XM_005251247.2
XP_005251304.1
2
416
XM_005251248.1
XP_005251305.1
2
102
XM_005251250.2
XP_005251307.1
2
416
XM_017013419.1
XP_016868908.1
2
416
XM_017013420.1
XP_016868909.1
2
416
XM_017013421.1
XP_016868910.1
2
416
XM_017013422.1
XP_016868911.1
2
416
XM_017013423.1
XP_016868912.1
2
416
XM_017013424.1
XP_016868913.1
2
416
XM_017013425.1
XP_016868914.1
2
416
XM_017013426.1
XP_016868915.1
2
194
XM_017013427.1
XP_016868916.1
2
194
XM_017013428.1
XP_016868917.1
2
416
XM_017013429.1
XP_016868918.1
2
416
XM_017013430.1
XP_016868919.1
2
416
XM_017013431.1
XP_016868920.1
2
102
XM_017013432.1
XP_016868921.1
2
416
XM_017013433.1
XP_016868922.1
2
102
XM_017013434.1
XP_016868923.1
2
416
XM_017013435.1
XP_016868924.1
2
194
XM_017013436.1
XP_016868925.1
2
416
XM_017013437.1
XP_016868926.1
2
416
XM_017013438.1
XP_016868927.1
2
416
XM_017013439.1
XP_016868928.1
2
102
XM_017013440.1
XP_016868929.1
2
194
XM_017013441.1
XP_016868930.1
2
416
XM_017013442.1
XP_016868931.1
2
416
XM_017013443.1
XP_016868932.1
2
416
XM_017013444.1
XP_016868933.1
2
102
XM_017013445.1
XP_016868934.1
2
102
XM_017013446.1
XP_016868935.1
2
102
XM_017013447.1
XP_016868936.1
2
102
GenBank mRNA
AF184241.1
-
2
132
AF224468.1
-
2
202
AF224469.1
-
2
177
AF289489.1
-
2
189
AF306765.1
-
2
173
AK291836.1
-
2
260
AK295528.1
-
2
216
AK298577.1
-
3
368
AK304314.1
-
2
208
BC015518.2
-
2
366
BC025236.1
-
2
375
BC066929.1
-
2
343
BC142967.1
-
2
324
BC144362.1
-
2
182
BC144363.1
-
2
183
CR996516.1
-
2
447
DA136318.1
-
2
416
DN993915.1
-
2
260
EU718494.1
-
2
255
FJ461473.1
-
2
178
S83325.1
-
2
189
U03109.1
-
2
255
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00188872_m1
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
pattern specification process
negative regulation of cell proliferation
regulation of protein stability
ion transmembrane transport
limb morphogenesis
peptidyl-aspartic acid hydroxylation
positive regulation of proteolysis
oxidation-reduction process
palate development
face morphogenesis
activation of cysteine-type endopeptidase activity
regulation of protein depolymerization
regulation of cardiac conduction
peptide-aspartate beta-dioxygenase activity
structural molecule activity
calcium ion binding
protein binding
structural constituent of muscle
electron carrier activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7fd786455c-crs9h:80/10.239.160.176:80.
git-commit: 5e88b81a8914dcee342b057544555f8ab0317e32
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.1-2024.10.42-1.0