Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CD207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015717.4 | NP_056532.4 | 3 | 464 | ||
XM_011532874.1 | XP_011531176.1 | 4 | 652 | |||
XM_011532875.2 | XP_011531177.1 | 5 | 669 | |||
XM_011532876.2 | XP_011531178.1 | 5 | 668 | |||
GenBank mRNA | AJ242859.1 | - | 3 | 464 | ||
AK314927.1 | - | 3 | 458 | |||
BC022278.1 | - | 3 | 457 | 4692155 |