Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020632.2 | NP_065683.2 | 11, 12 | 1309 | ||
NM_130840.2 | NP_570855.2 | 10, 11 | 1206 | |||
NM_130841.2 | NP_570856.2 | 10, 11 | 1264 | |||
XM_005250393.1 | XP_005250450.1 | 12, 13 | 1656 | |||
XM_005250394.3 | XP_005250451.1 | 10, 11 | 1568 | |||
GenBank mRNA | AF245517.1 | - | 11, 12 | 1294 | ||
AK055789.1 | - | 6, 7 | 697 | |||
AK292945.1 | - | 11, 12 | 1309 | |||
BC109304.1 | - | 10, 11 | 1114 | 40035236 | ||
BC109305.1 | - | 10, 11 | 1114 | 40035237 |