Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077196.1 | NP_001070664.1 | 15 | 1357 | ||
NM_001077197.1 | NP_001070665.1 | 19 | 1887 | |||
NM_001077358.1 | NP_001070826.1 | 17 | 1597 | |||
NM_016953.3 | NP_058649.3 | 18 | 2856 | |||
GenBank mRNA | AB036704.1 | - | 18 | 2856 | ||
AB038041.1 | - | 19 | 1887 | |||
AF281865.1 | - | 17 | 1569 | |||
AJ251509.1 | - | 15 | 1356 | |||
AJ278682.1 | - | 19 | 1930 | |||
AK074273.1 | - | 8 | 754 | |||
AK294076.1 | - | 3 | 145 | |||
BC112393.1 | - | 18 | 2847 | 40068843 | ||
BC114431.1 | - | 18 | 2847 | 40068833 | ||
HM771389.1 | - | 18 | 2538 | |||
HM771390.1 | - | 18 | 2538 | |||
HM771391.1 | - | 18 | 2538 |