Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016045.2 | NP_057129.2 | 3 | 336 | ||
GenBank mRNA | AF151865.1 | - | 3 | 304 | ||
AK002157.1 | - | 3 | 279 | |||
AK314449.1 | - | 3 | 276 | |||
BC010649.1 | - | 3 | 288 | 3873112 | ||
BC013969.2 | - | 3 | 302 | 3631601 | ||
CO248221.1 | - | 3 | 354 |
Gene Symbol: SLMO2-ATP5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037929.1 | 3 | 336 | |||
GenBank mRNA | FY212904.1 | - | 3 | 225 |