Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286077.1 | NP_001273006.1 | 3 | 372 | ||
NM_015853.4 | NP_056937.2 | 3 | 372 | |||
XM_005274033.4 | XP_005274090.1 | 4 | 285 | |||
XM_011545090.2 | XP_011543392.1 | 3 | 331 | |||
XM_017017874.1 | XP_016873363.1 | 4 | 281 | |||
GenBank mRNA | AK054832.1 | - | 2 | 449 | ||
AK297856.1 | - | 3 | 350 | |||
AK300542.1 | - | 3 | 328 | |||
AK316012.1 | - | 3 | 322 | |||
BC000902.2 | - | 3 | 311 | 2900493 | ||
BC001372.2 | - | 3 | 310 | 3051213 | ||
BC032689.1 | - | 2 | 434 | |||
BC040129.1 | - | 3 | 347 | 6065505 | ||
BP329601.1 | - | 3 | 372 | |||
M68864.1 | - | 3 | 313 |