Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015871.4 | NP_056955.2 | 2 | 326 | ||
XM_017001398.1 | XP_016856887.1 | 2 | 695 | |||
GenBank mRNA | AK311926.1 | - | 2 | 326 | ||
BC002580.2 | - | 2 | 304 | 3143091 | ||
BC019267.1 | - | 2 | 299 | 2823658 | ||
D45213.1 | - | 2 | 235 | |||
DA124660.1 | - | 2 | 326 |