Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252406.2 | NP_001239335.1 | 5 | 2052 | ||
NM_001256455.1 | NP_001243384.1 | 3 | 1812 | |||
NR_045515.1 | 6 | 1879 | ||||
NR_046206.1 | 5 | 1735 | ||||
NR_049765.1 | 4 | 1656 | ||||
XM_006711349.2 | XP_006711412.2 | 5 | 1775 | |||
XM_006711353.1 | XP_006711416.1 | 4 | 1630 | |||
XM_006711354.1 | XP_006711417.1 | 5 | 1718 | |||
XM_006711356.3 | XP_006711419.1 | 4 | 2129 | |||
XM_006711357.1 | XP_006711420.1 | 4 | 1968 | |||
XM_006711358.1 | XP_006711421.1 | 3 | 1819 | |||
XM_006711359.2 | XP_006711422.1 | 5 | 1781 | |||
XM_011509598.2 | XP_011507900.1 | 5 | 2410 | |||
XM_011509599.2 | XP_011507901.1 | 5 | 2388 | |||
XM_017001399.1 | XP_016856888.1 | 3 | 2051 | |||
GenBank mRNA | AF007833.1 | - | 2 | 1704 | ||
AK291619.1 | - | 5 | 1714 | |||
AK304762.1 | - | 5 | 1863 | |||
BC012070.1 | - | 4 | 1629 | 4552229 | ||
CR749303.1 | - | 6 | 1858 |