Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127713.1 | NP_001121185.1 | 8 | 1089 | ||
NM_015915.4 | NP_056999.2 | 7 | 905 | |||
NM_181598.3 | NP_853629.2 | 7 | 905 | |||
GenBank mRNA | AF131801.1 | - | 6 | 429 | ||
AF444143.1 | - | 7 | 836 | |||
AK223436.1 | - | 7 | 818 | |||
AK290185.1 | - | 7 | 818 | |||
AK312518.1 | - | 7 | 791 | |||
AL833591.1 | - | 8 | 873 | |||
AY032844.1 | - | 7 | 833 | |||
BC010708.2 | - | 7 | 808 | 3869877 | ||
CR457153.1 | - | 6 | 415 | |||
KU178621.1 | - | 7 | 664 | |||
KU178622.1 | - | 7 | 664 | |||
KU178623.1 | - | 7 | 664 |