Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015963.5 | NP_057047.4 | 2 | 442 | ||
XM_005247016.4 | XP_005247073.3 | 2 | 675 | |||
XM_011511291.2 | XP_011509593.1 | 2 | 244 | |||
XM_017004256.1 | XP_016859745.1 | 2 | 236 | |||
GenBank mRNA | AF258556.1 | - | 2 | 194 | ||
BC069235.1 | - | 2 | 372 | 6645198 | ||
BX405245.2 | - | 2 | 462 |