Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025780.2 | NP_001020951.1 | 4 | 1424 | ||
NM_016014.3 | NP_057098.2 | 4 | 1424 | |||
XM_006717134.3 | XP_006717197.2 | 4 | 875 | |||
XM_017014787.1 | XP_016870276.1 | 4 | 1031 | |||
XM_017014788.1 | XP_016870277.1 | 5 | 1816 | |||
XM_017014789.1 | XP_016870278.1 | 4 | 962 | |||
XM_017014790.1 | XP_016870279.1 | 3 | 1232 | |||
XM_017014791.1 | XP_016870280.1 | 3 | 899 | |||
GenBank mRNA | AF151825.1 | - | 4 | 1108 | ||
AK293060.1 | - | 4 | 1424 | |||
BC038390.1 | - | 4 | 921 | 4748883 | ||
BC044576.1 | - | 4 | 1372 | 5285591 |