Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RMDN1 SIRNA Assays ›
Gene Symbol:
RMDN1
Gene Name:
regulator of microtubule dynamics 1
Gene Aliases:
CGI-90, FAM82B, RMD-1, RMD1
Chromosome Location:
Chr.8: 86467116 - 86514348 on Build GRCh38
UniGene ID:
Hs.145386
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27472
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RMDN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286707.1
NP_001273636.1
6
736
NM_001286719.1
NP_001273648.1
6
736
NM_001317807.1
NP_001304736.1
7
644
NM_016033.2
NP_057117.2
7
826
XM_005250944.3
XP_005251001.1
7
732
XM_005250947.3
XP_005251004.1
6
642
XM_005250948.3
XP_005251005.1
7
634
XM_006716573.3
XP_006716636.1
7
821
XM_011517090.2
XP_011515392.1
7
806
XM_011517091.2
XP_011515393.1
7
806
XM_011517092.2
XP_011515394.1
6
716
XM_011517093.2
XP_011515395.1
7
806
XM_011517094.2
XP_011515396.1
7
806
XM_011517095.2
XP_011515397.1
7
806
XM_011517096.2
XP_011515398.1
7
806
XM_011517097.2
XP_011515399.1
6
716
XM_011517098.2
XP_011515400.1
6
716
XM_017013518.1
XP_016869007.1
7
732
XM_017013519.1
XP_016869008.1
7
806
XM_017013520.1
XP_016869009.1
7
732
XM_017013521.1
XP_016869010.1
7
732
XM_017013522.1
XP_016869011.1
7
806
XM_017013523.1
XP_016869012.1
7
732
XM_017013524.1
XP_016869013.1
7
634
XM_017013525.1
XP_016869014.1
7
634
XM_017013526.1
XP_016869015.1
7
634
XM_017013527.1
XP_016869016.1
6
731
XM_017013528.1
XP_016869017.1
6
539
GenBank mRNA
AF151848.1
-
7
738
AK000672.1
-
7
734
AK297896.1
-
6
637
AK303122.1
-
6
588
BC008509.1
-
5
390
BC009671.1
-
7
718
3896168
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0