Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRAK4 SIRNA Assays ›
Gene Symbol:
IRAK4
Gene Name:
interleukin 1 receptor associated kinase 4
Gene Aliases:
IPD1, IRAK-4, NY-REN-64, REN64
Chromosome Location:
Chr.12: 43758913 - 43789543 on Build GRCh38
UniGene ID:
Hs.138499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27528
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRAK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114182.2
NP_001107654.1
8
933
NM_001145256.1
NP_001138728.1
7
787
NM_001145257.1
NP_001138729.1
6
739
NM_001145258.1
NP_001138730.1
5
569
NM_016123.3
NP_057207.2
7
885
XM_005268943.3
XP_005269000.1
7
990
XM_005268944.4
XP_005269001.1
7
954
XM_005268945.4
XP_005269002.1
7
1198
XM_005268947.4
XP_005269004.1
8
944
XM_005268948.2
XP_005269005.1
9
984
XM_005268949.2
XP_005269006.1
6
844
XM_006719438.3
XP_006719501.1
8
1246
XM_006719439.2
XP_006719502.1
7
1351
XM_006719440.2
XP_006719503.1
7
762
XM_006719441.2
XP_006719504.1
6
647
XM_006719442.2
XP_006719505.1
7
651
XM_006719443.3
XP_006719506.1
6
794
XM_011538431.2
XP_011536733.1
8
1002
XM_011538432.1
XP_011536734.1
8
1038
XM_011538433.2
XP_011536735.1
7
1617
XM_017019390.1
XP_016874879.1
9
1119
XM_017019391.1
XP_016874880.1
8
1084
GenBank mRNA
AF155118.1
-
7
852
AF445802.1
-
6
803
AK000528.1
-
7
832
AK299944.1
-
6
736
AK304583.1
-
5
572
AK314288.1
-
7
874
AY283670.1
-
6
803
AY283671.1
-
6
801
AY340962.1
-
7
868
AY340963.1
-
8
916
AY340964.1
-
6
722
AY340965.1
-
7
770
AY340966.1
-
8
935
AY340967.1
-
9
983
AY730682.1
-
6
803
BC013316.1
-
7
873
4287014
KU178631.1
-
6
803
KU178632.1
-
5
657
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
toll-like receptor signaling pathway
neutrophil mediated immunity
MyD88-dependent toll-like receptor signaling pathway
JNK cascade
cytokine-mediated signaling pathway
toll-like receptor 9 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of smooth muscle cell proliferation
neutrophil migration
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
interleukin-1 receptor binding
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0