Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193609.1 | NP_001180538.1 | 2 | 410 | ||
NM_001193610.1 | NP_001180539.1 | 2 | 410 | |||
NM_001193611.1 | NP_001180540.1 | 2 | 410 | |||
NM_001193612.1 | NP_001180541.1 | 2 | 410 | |||
NM_016261.3 | NP_057345.2 | 2 | 410 | |||
XM_011524871.2 | XP_011523173.1 | 2 | 408 | |||
XM_017024716.1 | XP_016880205.1 | 2 | 408 | |||
XM_017024717.1 | XP_016880206.1 | 2 | 413 | |||
XM_017024719.1 | XP_016880208.1 | 2 | 413 | |||
GenBank mRNA | AB030837.1 | - | 1 | 123 | ||
AB031044.1 | - | 1 | 123 | |||
AB031045.1 | - | 1 | 123 | |||
AF201333.1 | - | 1 | 122 | |||
AK022771.1 | - | 3 | 300 | |||
AK024096.1 | - | 2 | 182 | |||
AK298492.1 | - | 2 | 323 | |||
AK315172.1 | - | 2 | 406 | |||
BC000258.1 | - | 2 | 385 | 3358263 | ||
BG682935.1 | - | 2 | 412 | |||
DA607129.1 | - | 2 | 400 |