Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083899.2 | NP_001077368.2 | 8 | 1870 | ||
NM_001256017.2 | NP_001242946.2 | 7 | 1812 | |||
NM_016363.5 | NP_057447.5 | 8 | 1866 | |||
GenBank mRNA | AB043819.1 | - | 8 | 1866 | ||
AB043820.1 | - | 7 | 1812 | |||
AB043821.1 | - | 8 | 1870 |