Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297748.1 | NP_001284677.1 | 3 | 544 | ||
NM_001297749.1 | NP_001284678.1 | 2 | 367 | |||
NM_001297750.1 | NP_001284679.1 | 5 | 741 | |||
NM_001297751.1 | NP_001284680.1 | 4 | 685 | |||
NM_016511.3 | NP_057595.2 | 4 | 643 | |||
XM_011520687.2 | XP_011518989.1 | 5 | 724 | |||
GenBank mRNA | AF200949.1 | - | 4 | 581 | ||
AK075114.1 | - | 4 | 563 | |||
AK091989.1 | - | 3 | 495 | |||
AK291584.1 | - | 4 | 531 | |||
AK300979.1 | - | 2 | 254 | |||
AK313589.1 | - | 4 | 561 | |||
AY358587.1 | - | 4 | 643 | |||
BC039072.1 | - | 4 | 549 | 5178017 | ||
BC067746.1 | - | 4 | 563 | 30337675 | ||
BM920364.1 | - | 584 | ||||
BQ722745.1 | - | 4 | 572 | |||
BX339075.2 | - | 4 | 581 | |||
BX380212.2 | - | 5 | 598 |