Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297748.1 | NP_001284677.1 | 4, 5 | 705 | ||
NM_001297749.1 | NP_001284678.1 | 3, 4 | 528 | |||
NM_001297750.1 | NP_001284679.1 | 6, 7 | 902 | |||
NM_001297751.1 | NP_001284680.1 | 5, 6 | 846 | |||
NM_016511.3 | NP_057595.2 | 5, 6 | 804 | |||
XM_011520687.2 | XP_011518989.1 | 6, 7 | 885 | |||
GenBank mRNA | AF200949.1 | - | 5, 6 | 742 | ||
AK075114.1 | - | 5, 6 | 724 | |||
AK091989.1 | - | 4, 5 | 656 | |||
AK291584.1 | - | 5, 6 | 692 | |||
AK300979.1 | - | 3, 4 | 415 | |||
AK313589.1 | - | 5, 6 | 722 | |||
AY358587.1 | - | 5, 6 | 804 | |||
BC039072.1 | - | 5, 6 | 710 | 5178017 | ||
BC067746.1 | - | 5, 6 | 724 | 30337675 | ||
BQ722745.1 | - | 5, 6 | 733 | |||
BX339075.2 | - | 5, 6 | 742 | |||
BX380212.2 | - | 6, 7 | 759 |