Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001795.4 | NP_001786.2 | 4, 5 | 795 | ||
XM_011522801.1 | XP_011521103.1 | 4, 5 | 772 | |||
GenBank mRNA | AB035304.1 | - | 4, 5 | 692 | ||
AB209908.1 | - | 4, 5 | 695 | |||
AK300461.1 | - | 4, 5 | 617 | |||
BC096363.3 | - | 4, 5 | 685 | 40012042 | ||
BC096364.3 | - | 4, 5 | 685 | 40012045 | ||
BC096365.1 | - | 4, 5 | 685 | 40012047 | ||
BC117520.2 | - | 4, 5 | 685 | 40012053 | ||
U84722.1 | - | 4, 5 | 743 | |||
X59796.1 | - | 4, 5 | 715 | |||
X79981.1 | - | 3, 4 | 648 |