Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016548.3 | NP_057632.2 | 3 | 504 | ||
NM_177937.2 | NP_808800.1 | 3 | 496 | |||
GenBank mRNA | AF236056.1 | - | 3 | 445 | ||
AK027717.1 | - | 3 | 467 | |||
AK075542.1 | - | 3 | 438 | |||
AK075557.1 | - | 3 | 408 | |||
AK092943.1 | - | 4 | 384 | |||
AY358593.1 | - | 3 | 409 | |||
BC001740.1 | - | 3 | 419 | 3351316 | ||
BQ216766.1 | - | 463 | ||||
CR457201.1 | - | 2 | 295 | |||
DA116177.1 | - | 3 | 497 | |||
EU794616.1 | - | 3 | 463 |