Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016548.3 | NP_057632.2 | 7 | 954 | ||
NM_177937.2 | NP_808800.1 | 7 | 946 | |||
GenBank mRNA | AF236056.1 | - | 7 | 895 | ||
AK027717.1 | - | 7 | 917 | |||
AK075542.1 | - | 7 | 888 | |||
AK075557.1 | - | 7 | 858 | |||
AK092943.1 | - | 8 | 834 | |||
AY358593.1 | - | 7 | 859 | |||
BC001740.1 | - | 7 | 869 | 3351316 | ||
EU794616.1 | - | 7 | 913 |