Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016565.2 | NP_057649.2 | 2 | 298 | ||
XM_017017883.1 | XP_016873372.1 | 2 | 614 | |||
XM_017017884.1 | XP_016873373.1 | 3 | 451 | |||
GenBank mRNA | AF242180.1 | - | 1 | 351 | ||
AK000434.1 | - | 2 | 298 | |||
AK314104.1 | - | 2 | 614 | |||
AL359618.1 | - | 2 | 278 | |||
BC106036.1 | - | 2 | 314 | 4445548 |