Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEX3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016626.4 | NP_057710.3 | 2 | 1937 | ||
GenBank mRNA | AK096190.1 | - | 2 | 1178 | ||
AL833519.1 | - | 1 | 797 | |||
AY950679.1 | - | 2 | 1576 | |||
BC041122.1 | - | 1 | 228 | |||
BC153820.1 | - | 3 | 983 | |||
KC456343.1 | - | 3 | 1082 | |||
KC456344.1 | - | 3 | 1066 |