Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033569.1 | NP_001028741.1 | 7 | 1011 | ||
NM_001033570.1 | NP_001028742.1 | 7 | 1007 | |||
NM_001033571.1 | NP_001028743.1 | 7 | 983 | |||
NM_001033572.1 | NP_001028744.1 | 7 | 979 | |||
NM_001033574.1 | NP_001028746.1 | 5 | 1390 | |||
NM_001289054.1 | NP_001275983.1 | 7 | 991 | |||
NM_001289056.1 | NP_001275985.1 | 7 | 1082 | |||
NM_016627.4 | NP_057711.3 | 6 | 1564 | |||
XM_005257432.4 | XP_005257489.1 | 6 | 1615 | |||
XM_005257433.3 | XP_005257490.1 | 7 | 1013 | |||
XM_005257435.4 | XP_005257492.1 | 7 | 955 | |||
XM_005257436.3 | XP_005257493.1 | 7 | 1342 | |||
XM_006721940.2 | XP_006722003.1 | 7 | 953 | |||
XM_017024723.1 | XP_016880212.1 | 7 | 1202 | |||
XM_017024724.1 | XP_016880213.1 | 7 | 951 | |||
XM_017024725.1 | XP_016880214.1 | 6 | 1527 | |||
XM_017024726.1 | XP_016880215.1 | 7 | 988 | |||
XM_017024727.1 | XP_016880216.1 | 7 | 989 | |||
XM_017024728.1 | XP_016880217.1 | 6 | 1056 | |||
GenBank mRNA | AJ635358.1 | - | 5 | 862 | ||
AK090981.1 | - | 5 | 1850 | |||
AK126146.1 | - | 6 | 1994 | |||
BC050709.1 | - | 7 | 943 | 6060235 | ||
BC056271.1 | - | 7 | 976 | 4102397 |