Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243266.1 | NP_001230195.1 | 3 | 405 | ||
NM_024021.3 | NP_076926.2 | 4 | 551 | |||
NM_148975.2 | NP_683876.1 | 3 | 405 | |||
XM_017017909.1 | XP_016873398.1 | 3 | 418 | |||
GenBank mRNA | AB013102.1 | - | 3 | 246 | ||
AB022821.1 | - | 3 | 270 | |||
AF068288.1 | - | 3 | 339 | |||
AF237912.1 | - | 3 | 356 | |||
AF350500.1 | - | 2 | 309 | |||
AF354928.1 | - | 2 | 229 | |||
AK075081.1 | - | 4 | 436 | |||
AK292089.1 | - | 3 | 360 | |||
AK314033.1 | - | 3 | 248 | |||
BC020648.1 | - | 3 | 280 | 4720088 | ||
BG538851.1 | - | 3 | 405 | |||
CB046393.1 | - | 3 | 274 |