Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015891.2 | NP_056975.1 | 10, 11 | 1122 | ||
GenBank mRNA | AB208903.1 | - | 10, 11 | 1096 | ||
AF038392.1 | - | 10, 11 | 1122 | |||
AF061241.1 | - | 10, 11 | 1095 | |||
AK222721.1 | - | 10, 11 | 1119 | |||
AK314601.1 | - | 10, 11 | 1118 | |||
BC111061.1 | - | 10, 11 | 2528 | |||
BC117145.1 | - | 10, 11 | 1119 | 40125696 | ||
BC126114.1 | - | 10, 11 | 1119 | 8991830 |