Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129884.2 | NP_001123356.1 | 4 | 501 | ||
NM_001243725.1 | NP_001230654.1 | 3 | 609 | |||
NM_001243726.1 | NP_001230655.1 | 3 | 393 | |||
NM_001278195.1 | NP_001265124.1 | 4 | 541 | |||
NM_015926.5 | NP_057010.1 | 3 | 449 | |||
NR_024012.2 | 2 | 157 | ||||
NR_103462.1 | 2 | 120 | ||||
NR_103463.1 | 2 | 89 | ||||
XM_006713195.3 | XP_006713258.1 | 3 | 389 | |||
XM_006713197.3 | XP_006713260.1 | 3 | 356 | |||
XM_006713198.2 | XP_006713261.1 | 3 | 424 | |||
XM_011533805.2 | XP_011532107.1 | 3 | 1104 | |||
XM_017006573.1 | XP_016862062.1 | 3 | 609 | |||
XM_017006574.1 | XP_016862063.1 | 2 | 335 | |||
GenBank mRNA | AF072733.1 | - | 2 | 347 | ||
AK023997.1 | - | 3 | 392 | |||
AK222938.1 | - | 3 | 361 | |||
AK222949.1 | - | 3 | 361 | |||
AL832608.1 | - | 1 | 2998 | |||
AY358972.1 | - | 2 | 348 | |||
BC001592.1 | - | 3 | 371 | |||
BC008742.2 | - | 3 | 609 | 3346733 | ||
BG772477.1 | - | 2 | 162 | |||
BI826172.1 | - | 4 | 461 | |||
BP252919.1 | - | 4 | 431 | |||
CB142778.1 | - | 2 | 121 | |||
DA894396.1 | - | 3 | 418 |