Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRAID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170795.1 | NP_001164266.1 | 5 | 464 | ||
NM_016085.4 | NP_057169.2 | 5 | 604 | |||
NM_080592.3 | NP_542159.3 | 5 | 784 | |||
GenBank mRNA | AB009017.1 | - | 5 | 477 | ||
AF077037.1 | - | 5 | 466 | |||
AF144055.2 | - | 5 | 594 | |||
AF275744.1 | - | 6 | 458 | |||
AK291894.1 | - | 5 | 594 | |||
AY358968.1 | - | 5 | 457 | |||
BC002846.1 | - | 5 | 452 | |||
BC011006.1 | - | 5 | 458 | |||
BC021237.2 | - | 5 | 564 | 4303513 | ||
BC035850.1 | - | 5 | 450 | 5753421 | ||
BI194121.1 | - | 5 | 605 | |||
BQ420456.1 | - | 5 | 587 | |||
BQ718371.1 | - | 5 | 499 | |||
CF126865.1 | - | 5 | 597 |