Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220488.1 | NP_001207417.1 | 10 | 1540 | ||
NM_001220489.1 | NP_001207418.1 | 8 | 1317 | |||
NM_001220490.1 | NP_001207419.1 | 8 | 1225 | |||
NM_001257.4 | NP_001248.1 | 9 | 1434 | |||
XM_011522804.2 | XP_011521106.1 | 8 | 1041 | |||
GenBank mRNA | AK294277.1 | - | 3 | 297 | ||
AK295886.1 | - | 8 | 1055 | |||
AK298612.1 | - | 8 | 1143 | |||
AK304733.1 | - | 10 | 1295 | |||
AK307862.1 | - | 10 | 1232 | |||
BC028624.2 | - | 9 | 1226 | 4825734 | ||
BC030653.2 | - | 9 | 1226 | 4811561 | ||
BU162665.1 | - | 5 | 567 | |||
CN263227.1 | - | 4 | 458 | |||
DA561809.1 | - | 2 | 110 | |||
DB003683.1 | - | 4 | 507 | |||
L34058.1 | - | 9 | 1579 | |||
U59288.1 | - | 9 | 1351 |