Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPHLN1 SIRNA Assays ›
Gene Symbol:
PPHLN1
Gene Name:
periphilin 1
Gene Aliases:
CR, HSPC206, HSPC232
Chromosome Location:
Chr.12: 42326126 - 42448621 on Build GRCh38
UniGene ID:
Hs.444157
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s28288
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPHLN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143787.1
NP_001137259.1
4
277
NM_001143788.1
NP_001137260.1
5
442
NM_001143789.1
NP_001137261.1
4
277
NM_016488.6
NP_057572.5
5
442
NM_201438.1
NP_958846.1
5
320
NM_201439.1
NP_958847.1
5
442
NM_201440.1
NP_958848.1
5
320
NM_201515.1
NP_958923.1
6
485
XM_005268962.3
XP_005269019.1
4
296
XM_011538459.2
XP_011536761.1
6
463
XM_011538465.2
XP_011536767.1
5
420
XM_011538467.2
XP_011536769.1
5
302
XM_011538470.2
XP_011536772.1
5
320
XM_011538471.2
XP_011536773.1
4
296
XM_017019434.1
XP_016874923.1
6
463
XM_017019435.1
XP_016874924.1
6
462
XM_017019436.1
XP_016874925.1
6
463
XM_017019437.1
XP_016874926.1
5
420
XM_017019438.1
XP_016874927.1
5
420
XM_017019439.1
XP_016874928.1
6
463
XM_017019440.1
XP_016874929.1
6
463
XM_017019441.1
XP_016874930.1
5
420
XM_017019442.1
XP_016874931.1
5
420
XM_017019443.1
XP_016874932.1
6
463
XM_017019444.1
XP_016874933.1
5
302
XM_017019445.1
XP_016874934.1
4
296
XM_017019446.1
XP_016874935.1
4
295
XM_017019447.1
XP_016874936.1
5
320
XM_017019448.1
XP_016874937.1
5
320
XM_017019449.1
XP_016874938.1
4
296
XM_017019450.1
XP_016874939.1
4
296
XM_017019451.1
XP_016874940.1
5
320
XM_017019452.1
XP_016874941.1
6
463
XM_017019453.1
XP_016874942.1
5
320
GenBank mRNA
AF151040.1
-
5
284
AF151066.1
-
5
295
AK055690.1
-
5
430
AK129910.1
-
6
485
AK299951.1
-
4
259
AK303612.1
-
4
277
AK303680.1
-
3
215
AK308022.1
-
5
416
AY039238.1
-
5
424
AY157850.1
-
6
481
BC025306.1
-
4
244
BC039832.1
-
5
287
6051282
BC040449.1
-
5
287
CF242917.1
-
4
292
DC424443.1
-
4
262
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
keratinization
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0