Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143780.2 | NP_001137252.1 | 12 | 1199 | ||
NM_001321240.1 | NP_001308169.1 | 11 | 1130 | |||
NM_001321241.1 | NP_001308170.1 | 12 | 1193 | |||
NM_016016.3 | NP_057100.1 | 12 | 1175 | |||
XM_011524880.2 | XP_011523182.1 | 12 | 1174 | |||
XM_011524882.2 | XP_011523184.1 | 11 | 1114 | |||
XM_011524883.2 | XP_011523185.1 | 11 | 1116 | |||
XM_011524884.1 | XP_011523186.1 | 11 | 1100 | |||
XM_017024737.1 | XP_016880226.1 | 11 | 1082 | |||
GenBank mRNA | AB209576.1 | - | 10 | 1787 | ||
AF119864.1 | - | 11 | 961 | |||
AF151827.1 | - | 12 | 1117 | |||
AF317711.1 | - | 11 | 1037 | |||
AK026060.1 | - | 12 | 1117 | |||
AK289357.1 | - | 12 | 1160 | |||
AK294085.1 | - | 11 | 1115 | |||
AL133584.1 | - | 12 | 1117 | |||
BC001398.2 | - | 12 | 1121 | 3051436 | ||
BC009330.2 | - | 12 | 1093 | 4126268 | ||
BC096819.1 | - | 12 | 1102 | 6109001 |