Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143780.2 | NP_001137252.1 | 4 | 354 | ||
NM_001321241.1 | NP_001308170.1 | 4 | 354 | |||
NM_016016.3 | NP_057100.1 | 4 | 354 | |||
XM_011524880.2 | XP_011523182.1 | 4 | 311 | |||
XM_011524883.2 | XP_011523185.1 | 4 | 321 | |||
XM_011524884.1 | XP_011523186.1 | 4 | 329 | |||
XM_017024737.1 | XP_016880226.1 | 4 | 329 | |||
GenBank mRNA | AB209576.1 | - | 3 | 764 | ||
AF119864.1 | - | 3 | 140 | |||
AF151827.1 | - | 4 | 296 | |||
AF317711.1 | - | 3 | 192 | |||
AK026060.1 | - | 4 | 296 | |||
AK289357.1 | - | 4 | 315 | |||
AK295478.1 | - | 4 | 315 | |||
AL133584.1 | - | 4 | 296 | |||
BC001398.2 | - | 4 | 300 | 3051436 | ||
BC009330.2 | - | 4 | 272 | 4126268 | ||
BC096819.1 | - | 4 | 257 | 6109001 | ||
CB960224.1 | - | 4 | 381 | |||
CV573041.1 | - | 4 | 333 |