- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Gene Symbol: C14orf166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016039.2 | NP_057123.1 | 4 | 487 | ||
GenBank mRNA | AF087877.1 | - | 4 | 518 | ||
AF100755.1 | - | 4 | 518 | |||
AF151857.1 | - | 4 | 518 | |||
AK312004.1 | - | 4 | 451 | |||
AY817175.1 | - | 4 | 357 | |||
BC001722.1 | - | 4 | 439 | 3528725 | ||
DQ525179.1 | - | 4 | 522 |