Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016071.3 | NP_057155.1 | 3 | 401 | ||
NM_053035.2 | NP_444263.1 | 3 | 343 | |||
GenBank mRNA | AF078858.1 | - | 3 | 317 | ||
AF151897.1 | - | 3 | 379 | |||
AK024776.1 | - | 3 | 399 | |||
BC015462.2 | - | 2 | 274 | 3889203 | ||
BC105793.1 | - | 3 | 312 | |||
BQ420818.1 | - | 3 | 343 |