Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 4 | 1179 | ||
NM_001293214.1 | NP_001280143.1 | 3 | 963 | |||
NM_001293215.1 | NP_001280144.1 | 4 | 1082 | |||
NM_001293216.1 | NP_001280145.1 | 4 | 981 | |||
NM_178014.3 | NP_821133.1 | 4 | 1095 | |||
GenBank mRNA | AB062393.1 | - | 4 | 921 | ||
AF070561.1 | - | 4 | 916 | |||
AF070593.1 | - | 4 | 916 | |||
AF070600.1 | - | 4 | 933 | |||
AF141349.1 | - | 4 | 933 | |||
AK297638.1 | - | 4 | 563 | |||
AK298889.1 | - | 4 | 985 | |||
AK302195.1 | - | 3 | 425 | |||
AK302314.1 | - | 4 | 1048 | |||
AK303227.1 | - | 3 | 682 | |||
AK309684.1 | - | 2 | 537 | |||
AK316265.1 | - | 4 | 852 | |||
BC001896.1 | - | 1 | 477 | |||
BC001938.1 | - | 4 | 920 | 2820585 | ||
BC002347.2 | - | 4 | 925 | 2964461 | ||
BC005838.2 | - | 4 | 920 | 2820585 | ||
BC007605.1 | - | 4 | 925 | 3350604 | ||
BC013374.2 | - | 4 | 916 | 3946253 | ||
BC015889.1 | - | 1 | 257 | |||
BC019924.1 | - | 4 | 913 | 4998571 | ||
BC020946.1 | - | 4 | 954 | 4795692 | ||
BC021909.1 | - | 4 | 915 | 3509417 | ||
BC029323.1 | - | 1 | 278 | |||
BC070326.1 | - | 4 | 931 | 6698679 | ||
BC103746.1 | - | 4 | 938 | 6503977 | ||
DQ457058.1 | - | 1 | 351 | |||
V00599.1 | - | 3 | 768 |