Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001803.2 | NP_001794.2 | 2 | 225 | ||
GenBank mRNA | A23013.1 | - | 2 | 160 | ||
FJ460513.1 | - | 2 | 225 | |||
KP176645.1 | - | 2 | 127 | |||
X62466.1 | - | 2 | 160 |