Gene Symbol: CD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001803.2 | NP_001794.2 | 1, 2 | 154 | ||
GenBank mRNA | A23013.1 | - | 1, 2 | 89 | ||
BC000644.2 | - | 1, 2 | 98 | 3345325 | ||
BI835519.1 | - | 1, 2 | 156 | |||
BT007116.1 | - | 1, 2 | 56 | |||
CR456874.1 | - | 1, 2 | 56 | |||
FJ460513.1 | - | 1, 2 | 154 | |||
KP176645.1 | - | 1, 2 | 56 | |||
X62466.1 | - | 1, 2 | 89 | |||
X67699.1 | - | 1, 2 | 80 |