Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001895.2 | NP_001001895.1 | 6 | 820 | ||
NM_001243467.1 | NP_001230396.1 | 6 | 820 | |||
NM_018961.3 | NP_061834.1 | 7 | 934 | |||
XM_006724013.3 | XP_006724076.1 | 7 | 1018 | |||
XM_011529605.2 | XP_011527907.1 | 7 | 934 | |||
XM_011529606.2 | XP_011527908.1 | 7 | 903 | |||
XM_011529607.2 | XP_011527909.1 | 7 | 987 | |||
XM_011529609.1 | XP_011527911.1 | 7 | 934 | |||
XM_011529610.2 | XP_011527912.1 | 3 | 2933 | |||
GenBank mRNA | AF520809.1 | - | 7 | 861 | ||
AF521702.1 | - | 6 | 747 | |||
AJ277750.1 | - | 7 | 881 | |||
AK308151.1 | - | 6 | 760 | |||
AK313367.1 | - | 6 | 801 | |||
BC028138.1 | - | 6 | 772 | 5215324 | ||
BC069357.1 | - | 7 | 881 | 7262315 | ||
BC069511.1 | - | 6 | 765 | 7262403 | ||
BC069577.1 | - | 6 | 767 |