Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL20RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278722.1 | NP_001265651.1 | 7 | 1228 | ||
NM_001278723.1 | NP_001265652.1 | 6 | 1368 | |||
NM_001278724.1 | NP_001265653.1 | 6 | 1302 | |||
NM_014432.3 | NP_055247.3 | 7 | 1547 | |||
XM_006715506.2 | XP_006715569.1 | 6 | 944 | |||
XM_011535904.2 | XP_011534206.1 | 6 | 1055 | |||
XM_017010954.1 | XP_016866443.1 | 7 | 1736 | |||
XM_017010955.1 | XP_016866444.1 | 7 | 1196 | |||
GenBank mRNA | AF184971.1 | - | 7 | 1331 | ||
AJ420490.1 | - | 3 | 522 | |||
AK098312.1 | - | 3 | 1137 | |||
AK297121.1 | - | 7 | 1190 | |||
AK315293.1 | - | 7 | 1335 | |||
AY358883.1 | - | 6 | 882 | |||
BC113574.1 | - | 7 | 1425 | 8322626 | ||
BC113602.1 | - | 7 | 1425 | 8322654 |