Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145944.1 | NP_001139416.1 | 10 | 1794 | ||
NM_001145945.1 | NP_001139417.1 | 12 | 1930 | |||
NM_001145946.1 | NP_001139418.1 | 13 | 2077 | |||
NM_017435.4 | NP_059131.1 | 12 | 2090 | |||
XM_005253394.2 | XP_005253451.1 | 13 | 1866 | |||
XM_005253396.2 | XP_005253453.1 | 11 | 6631 | |||
XM_011520703.2 | XP_011519005.1 | 13 | 1866 | |||
XM_011520704.2 | XP_011519006.1 | 13 | 1866 | |||
XM_011520709.2 | XP_011519011.1 | 10 | 1508 | |||
XM_011520711.2 | XP_011519013.1 | 12 | 6686 | |||
XM_017019484.1 | XP_016874973.1 | 15 | 7127 | |||
XM_017019485.1 | XP_016874974.1 | 14 | 7045 | |||
XM_017019486.1 | XP_016874975.1 | 12 | 6785 | |||
XM_017019487.1 | XP_016874976.1 | 11 | 6631 | |||
XM_017019488.1 | XP_016874977.1 | 16 | 7182 | |||
XM_017019489.1 | XP_016874978.1 | 17 | 7282 | |||
XM_017019490.1 | XP_016874979.1 | 17 | 7246 | |||
GenBank mRNA | AF205076.1 | - | 12 | 2091 | ||
AF260704.1 | - | 12 | 1786 | |||
AK294333.1 | - | 10 | 1662 | |||
AK296236.1 | - | 12 | 1823 | |||
AK303713.1 | - | 13 | 1953 | |||
AK308729.1 | - | 11 | 1726 | |||
AL834209.1 | - | 12 | 4852 | |||
BC022461.2 | - | 12 | 1819 | 4801171 |